Learn More
Abnova™ Human EPHA2 (NM_004431.2, 585 a.a. - 976 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4677
Catalog No. 89-943-391
Add to cart
Description
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands. [provided by RefSeq]
Specifications
Specifications
NM_004431.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
73.691kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
ECK
EPHA2
30pmol/μg x min
Yes
0.505 ug/uL
Liquid
1969
EPHA2 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
EPHA2
Insect cell (Sf9) expression system
Insect
GST-His