Learn More
Abnova™ Human EPHA3 (NP_005224.2, 1 a.a. - 464; 1 a.a. - 216 a.a.) Full-length Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P5704
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands. Two alternatively spliced transcript variants have been described for this gene. [provided by RefSeq]
Specifications
NP_005224.2 | |
Liquid | |
73kDa | |
Baculovirus infected insect cell (Sf21) expression system | |
Loading 1 ug protein in SDS-PAGE | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETK/ETK1/HEK/HEK4/TYRO4 | |
EPHA3 | |
Baculovirus | |
GST | |
Liquid |
Functional Study, SDS-PAGE | |
2042 | |
EPHA3 (Human) Recombinant Protein | |
Glutathione sepharose chromatography | |
5 ug | |
RUO | |
EPHA3 | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000. Substrate : Blk/Lyntide. ATP: 100 μM. |
|
Recombinant | |
Baculovirus infected insect cell (Sf21) expression system | |
88% by SDS-PAGE/CBB staining. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.