Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPHA4 Full-length ORF (no Protein_acc, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002043P01S

 View more versions of this product

Catalog No. 89-943-405


Add to cart

Description

Description

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq]

Sequence: MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP
Specifications

Specifications

no protein_acc
Liquid
2043
EPHA4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP
RUO
EPHA4
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HEK8|SEK|TYRO1
EPHA4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit