Learn More
Abnova™ Human EPHA4 (NM_004438, 570 a.a. - 986 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4679
Catalog No. 89-943-403
Add to cart
Description
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq]
Specifications
Specifications
NM_004438
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
76.587kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
HEK8|SEK|TYRO1
EPHA4
19pmol/μg x min
Yes
0.628 ug/uL
Liquid
2043
EPHA4 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
EPHA4
Insect cell (Sf9) expression system
Insect
GST-His