Learn More
Abnova™ Human EPHA4 (NP_004430.3, 586 a.a. - 986 a.a.) Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5705
Catalog No. 89-943-404
Add to cart
Description
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq]
Specifications
Specifications
NP_004429.1
Liquid
2043
EPHA4 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HEK8|SEK|TYRO1
EPHA4
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Blk/Lyntide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
72kDa
Baculovirus infected insect cell (Sf21) expression system
89% by SDS-PAGE/CBB staining.
5 ug
RUO
EPHA4
Baculovirus infected insect cell (Sf21) expression system
Insect
GST