Learn More
Abnova™ Human EPHA5 (NM_004439.3, 595 a.a. - 1037 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P4680
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
NM_004439.3 | |
Functional Study, SDS-PAGE | |
2044 | |
EPHA5 (Human) Recombinant Protein | |
GST affinity chromatography | |
100 ug | |
RUO | |
EPHA5 | |
247pmol/μg x min | |
Recombinant | |
Insect cell (Sf9) expression system |
0.167 ug/uL | |
Liquid | |
80.683kDa | |
Insect cell (Sf9) expression system | |
2 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing | |
CEK7/EHK1/HEK7/TYRO4 | |
EPHA5 | |
Baculovirus | |
GST-His | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.