Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPHA5 (NP_004430.3, 662 a.a. - 948 a.a.) Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5706

Catalog No. 89-943-411


Add to cart

Description

Description

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Specifications

NP_004430.3
Liquid
2044
EPHA5 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CEK7|EHK1|HEK7|TYRO4
EPHA5
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Blk/Lyntide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
59kDa
Baculovirus infected insect cell (Sf21) expression system
89% by SDS-PAGE/CBB staining.
5 ug
RUO
EPHA5
Baculovirus infected insect cell (Sf21) expression system
Insect
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit