Learn More
Abnova™ Human EPHA6 (CAD38242.1, 683 a.a. - 1130 a.a.) Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5546
Catalog No. 89-943-414
Add to cart
Specifications
Specifications
CAD38242.1
Liquid
285220
EPHA6 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434C1418|EPA6|FLJ35246|PRO57066
EPHA6
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Blk/Lyntide. ATP: 100 uM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
77kDa
Baculovirus infected insect cell (Sf21) expression system
84% by SDS-PAGE/CBB staining
5 ug
RUO
EPHA6
Baculovirus infected insect cell (Sf21) expression system
Insect
GST