Learn More
Abnova™ Human EPHB1 (NM_004441.2, 565 a.a. - 984 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4682
Catalog No. 89-943-422
Add to cart
Description
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq]
Specifications
Specifications
NM_004441.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
76.906kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
ELK|EPHT2|FLJ37986|Hek6|NET
EPHB1
72pmol/μg x min
Yes
0.500 ug/uL
Liquid
2047
EPHB1 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
EPHB1
Insect cell (Sf9) expression system
Insect
GST-His