Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPHB1 (NP_003148.2, 1 a.a. - 841 a.a.) Full-length Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5708

Catalog No. 89-943-423


Add to cart

Description

Description

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq]

Specifications

Specifications

NP_004432.1
Liquid
2047
EPHB1 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELK|EPHT2|FLJ37986|Hek6|NET
EPHB1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Blk/Lyntide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
73kDa
Baculovirus infected insect cell (Sf21) expression system
68% by SDS-PAGE/CBB staining.
5 ug
RUO
EPHB1
Baculovirus infected insect cell (Sf21) expression system
Insect
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit