Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPHB2 (NM_004442, 570 a.a. - end) Partial Recombinant Protein with GST-tag expressed in Sf9 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P3858

Catalog No. 89-943-427


Add to cart

Description

Description

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq]

Specifications

Specifications

NM_004442
Liquid
2048
EPHB2 (Human) Recombinant Protein
>90% by densitometry
5 ug
RUO
EPHB2
Insect cell (Sf9) expression system
Insect
GST
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
66kDa
Insect cell (Sf9) expression system
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAPB|DRT|EPHT3|ERK|Hek5|MGC87492|PCBC|Tyro5
EPHB2
The specific activity was determined to be 39nmol /min/mg as per Activity Assay Protocol.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit