Learn More
Abnova™ Human EPHB3 (NM_004443, 585 a.a. - 998 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4684
Catalog No. 89-943-432
Add to cart
Description
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq]
Specifications
Specifications
NM_004443
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
80.148kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
ETK2|HEK2|TYRO6
EPHB3
75pmol/μg x min
Yes
0.258 ug/uL
Liquid
2049
EPHB3 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
EPHB3
Insect cell (Sf9) expression system
Insect
GST-His