Learn More
Abnova™ Human EPHB4 (NM_004444.2, 563 a.a. - 987 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4685
Catalog No. 89-943-438
Add to cart
Description
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene binds to ephrin-B2 and plays an essential role in vascular development. [provided by RefSeq]
Specifications
Specifications
NM_004444.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
76.794kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
HTK|MYK1|TYRO11
EPHB4
78pmol/μg x min
Yes
0.235 ug/uL
Liquid
2050
EPHB4 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
EPHB4
Insect cell (Sf9) expression system
Insect
GST-His