Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human EPM2AIP1 Full-length ORF (NP_055620.1, 1 a.a. - 607 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00009852P01S
Description
The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless. [provided by RefSeq]
Sequence: MWMTPKRSKMEVDEALVFRPEWTQRYLVVEPPEGDGALCLVCRRLIVATRERDVRRHYEAEHEYYERYVADGERAALVERLRQGDLPVASFTPEERAARAGLGLCRLLALKGRGWGEGDFVYQCMEVLLREVLPEHVSVLQGVDLSPDITRQRILSIDRNLRNQLFNRARDFKAYSLALDDQAFVAYENYLLVFIRGVGPELEVQEDLLTIINLTHHFSVGALMSAILESLQTAGLSLQRMVGLTTTHTLRMIGENSGLVSYMREKAVSPNCWNVIHYSGFLHLELLSSYDVDVNQIINTISEWIVLIKTRGVRRPEFQTLLTESESEHGERVNGRCLNNWLRRGKTLKLIFSLRKEMEAFLVSVGATTVHFSDKQWLCDFGFLVDIMEHLRELSEELRVSKVFAAAAFDHICTFEVKLNLFQRHIEEKNLTDFPALREVVDELKQQNKEDEKIFDPDRYQMVICRLQKEFERHFKDLRFIKKDLELFSNPFNFKPEYAPISVRVELTKLQANTNLWNEYRIKDLGQFYAGLSAESYPIIKGVACKVASLFDSNQICEKAFSYLTRNQHTLSQPLTDEHLQALFRVATTEMEPGWDDLVRERNESNPSpecifications
NP_055620.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
96.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11207/KIAA0766 | |
EPM2AIP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9852 | |
EPM2AIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWMTPKRSKMEVDEALVFRPEWTQRYLVVEPPEGDGALCLVCRRLIVATRERDVRRHYEAEHEYYERYVADGERAALVERLRQGDLPVASFTPEERAARAGLGLCRLLALKGRGWGEGDFVYQCMEVLLREVLPEHVSVLQGVDLSPDITRQRILSIDRNLRNQLFNRARDFKAYSLALDDQAFVAYENYLLVFIRGVGPELEVQEDLLTIINLTHHFSVGALMSAILESLQTAGLSLQRMVGLTTTHTLRMIGENSGLVSYMREKAVSPNCWNVIHYSGFLHLELLSSYDVDVNQIINTISEWIVLIKTRGVRRPEFQTLLTESESEHGERVNGRCLNNWLRRGKTLKLIFSLRKEMEAFLVSVGATTVHFSDKQWLCDFGFLVDIMEHLRELSEELRVSKVFAAAAFDHICTFEVKLNLFQRHIEEKNLTDFPALREVVDELKQQNKEDEKIFDPDRYQMVICRLQKEFERHFKDLRFIKKDLELFSNPFNFKPEYAPISVRVELTKLQANTNLWNEYRIKDLGQFYAGLSAESYPIIKGVACKVASLFDSNQICEKAFSYLTRNQHTLSQPLTDEHLQALFRVATTEMEPGWDDLVRERNESNP | |
RUO | |
EPM2AIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction