Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPM2AIP1 Full-length ORF (NP_055620.1, 1 a.a. - 607 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009852P01L

 View more versions of this product

Catalog No. 89-943-462


Add to cart

Description

Description

The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless. [provided by RefSeq]

Sequence: MWMTPKRSKMEVDEALVFRPEWTQRYLVVEPPEGDGALCLVCRRLIVATRERDVRRHYEAEHEYYERYVADGERAALVERLRQGDLPVASFTPEERAARAGLGLCRLLALKGRGWGEGDFVYQCMEVLLREVLPEHVSVLQGVDLSPDITRQRILSIDRNLRNQLFNRARDFKAYSLALDDQAFVAYENYLLVFIRGVGPELEVQEDLLTIINLTHHFSVGALMSAILESLQTAGLSLQRMVGLTTTHTLRMIGENSGLVSYMREKAVSPNCWNVIHYSGFLHLELLSSYDVDVNQIINTISEWIVLIKTRGVRRPEFQTLLTESESEHGERVNGRCLNNWLRRGKTLKLIFSLRKEMEAFLVSVGATTVHFSDKQWLCDFGFLVDIMEHLRELSEELRVSKVFAAAAFDHICTFEVKLNLFQRHIEEKNLTDFPALREVVDELKQQNKEDEKIFDPDRYQMVICRLQKEFERHFKDLRFIKKDLELFSNPFNFKPEYAPISVRVELTKLQANTNLWNEYRIKDLGQFYAGLSAESYPIIKGVACKVASLFDSNQICEKAFSYLTRNQHTLSQPLTDEHLQALFRVATTEMEPGWDDLVRERNESNP
Specifications

Specifications

NP_055620.1
Liquid
9852
EPM2AIP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWMTPKRSKMEVDEALVFRPEWTQRYLVVEPPEGDGALCLVCRRLIVATRERDVRRHYEAEHEYYERYVADGERAALVERLRQGDLPVASFTPEERAARAGLGLCRLLALKGRGWGEGDFVYQCMEVLLREVLPEHVSVLQGVDLSPDITRQRILSIDRNLRNQLFNRARDFKAYSLALDDQAFVAYENYLLVFIRGVGPELEVQEDLLTIINLTHHFSVGALMSAILESLQTAGLSLQRMVGLTTTHTLRMIGENSGLVSYMREKAVSPNCWNVIHYSGFLHLELLSSYDVDVNQIINTISEWIVLIKTRGVRRPEFQTLLTESESEHGERVNGRCLNNWLRRGKTLKLIFSLRKEMEAFLVSVGATTVHFSDKQWLCDFGFLVDIMEHLRELSEELRVSKVFAAAAFDHICTFEVKLNLFQRHIEEKNLTDFPALREVVDELKQQNKEDEKIFDPDRYQMVICRLQKEFERHFKDLRFIKKDLELFSNPFNFKPEYAPISVRVELTKLQANTNLWNEYRIKDLGQFYAGLSAESYPIIKGVACKVASLFDSNQICEKAFSYLTRNQHTLSQPLTDEHLQALFRVATTEMEPGWDDLVRERNESNP
RUO
EPM2AIP1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11207|KIAA0766
EPM2AIP1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit