Learn More
Abnova™ Human EPO (P01588) Full-length Recombinant Protein expressed in CHO cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4137
Catalog No. 89-943-476
Add to cart
Description
Description
This gene is a member of the EPO/TPO family and encodes a secreted, glycosylated cytokine composed of four alpha helical bundles. The protein is found in the plasma and regulates red cell production by promoting erythroid differentiation and initiating hemoglobin synthesis. This protein also has neuroprotective activity against a variety of potential brain injuries and antiapoptotic functions in several tissue types. [provided by RefSeq]