Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPS8 Full-length ORF (NP_004438.3, 1 a.a. - 822 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002059P01L

 View more versions of this product

Catalog No. 89-943-487


Add to cart

Description

Description

This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq]

Sequence: MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
Specifications

Specifications

NP_004438.3
Liquid
2059
EPS8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
RUO
EPS8
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPS8
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit