Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPS8L1 Full-length ORF (NP_631943.1, 1 a.a. - 551 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054869P01

Catalog No. 89-943-490


Add to cart

Description

Description

This gene encodes a protein that is related to epidermal growth factor receptor pathway substrate 8 (EPS8), a substrate for the epidermal growth factor receptor. The function of this protein is unknown. At least two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGRKAIVLAIANTSLAFPLCQHLVTFCLGEDDGVHTVEDASRKLAVMDSQGRVWAQEMLLRVSPDHVTLLDPASKEELESYPLGAIVRCDAVMPPGRSRSLLLLVCQEPERAQPDVHFFQGLRLGAELIREDIQGALHNYRSGRGERRAAALRATQEELQRDRSPAAETPPLQRRPSVRAVISTVERGAGRGRPQAKPIPEAEEAQRPEPVGTSSNADSASPDLGPRGPDLAVLQAEREVDILNHVFDDVESFVSRLQKSAEAARVLEHRERGRRSRRRAAGEGLLTLRAKPPSEAEYTDVLQKIKYAFSLLARLRGNIADPSSPELLHFLFGPLQMIVNTSGGPEFASSVRRPHLTSDAVALLRDNVTPRENELWTSLGDSWTRPGLELSPEEGPPYRPEFFSGWEPPVTDPQSRAWEDPVEKQLQHERRRRQVTQATQQGRGWEVRGRGRSAWPRLTRLSYFLQQSAPQVAVNGHRDLEPESEPQLESETAGKWVLCNYDFQARNSSELSVKQRDVLEVLEDKEKVSELEAVMEKQKKKVEGEVEMEVI
Specifications

Specifications

NP_631943.1
Liquid
54869
EPS8L1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGRKAIVLAIANTSLAFPLCQHLVTFCLGEDDGVHTVEDASRKLAVMDSQGRVWAQEMLLRVSPDHVTLLDPASKEELESYPLGAIVRCDAVMPPGRSRSLLLLVCQEPERAQPDVHFFQGLRLGAELIREDIQGALHNYRSGRGERRAAALRATQEELQRDRSPAAETPPLQRRPSVRAVISTVERGAGRGRPQAKPIPEAEEAQRPEPVGTSSNADSASPDLGPRGPDLAVLQAEREVDILNHVFDDVESFVSRLQKSAEAARVLEHRERGRRSRRRAAGEGLLTLRAKPPSEAEYTDVLQKIKYAFSLLARLRGNIADPSSPELLHFLFGPLQMIVNTSGGPEFASSVRRPHLTSDAVALLRDNVTPRENELWTSLGDSWTRPGLELSPEEGPPYRPEFFSGWEPPVTDPQSRAWEDPVEKQLQHERRRRQVTQATQQGRGWEVRGRGRSAWPRLTRLSYFLQQSAPQVAVNGHRDLEPESEPQLESETAGKWVLCNYDFQARNSSELSVKQRDVLEVLEDKEKVSELEAVMEKQKKKVEGEVEMEVI
RUO
EPS8L1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.1kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DRC3|EPS8R1|FLJ20258|MGC23164|MGC4642|PP10566
EPS8L1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit