Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPS8L2 Partial ORF (NP_073609, 615 a.a. - 714 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064787Q01S

 View more versions of this product

Catalog No. 89-943-492


Add to cart

Description

Description

This gene encodes a member of the EPS8 gene family. The encoded protein, like other members of the family, is thought to link growth factor stimulation to actin organization, generating functional redundancy in the pathways that regulate actin cytoskeletal remodeling. [provided by RefSeq]

Sequence: ERSQPVSQPLTYESGPDEVRAWLEAKAFSPRIVENLGILTGPQLFSLNKEELKKVCGEEGVRVYSQLTMQKAFLEKQQSGSELEELMNKFHSMNQRRGED
Specifications

Specifications

NP_073609
Liquid
64787
EPS8L2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPS8R2|FLJ16738|FLJ21935|FLJ22171|MGC126530|MGC3088
EPS8L2
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ERSQPVSQPLTYESGPDEVRAWLEAKAFSPRIVENLGILTGPQLFSLNKEELKKVCGEEGVRVYSQLTMQKAFLEKQQSGSELEELMNKFHSMNQRRGED
RUO
EPS8L2
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit