Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPS8L3 Full-length ORF (AAH12926.1, 1 a.a. - 594 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079574P01

Catalog No. 89-943-498


Add to cart

Description

Description

This gene encodes a protein that is related to epidermal growth factor receptor pathway substrate 8 (EPS8), a substrate for the epidermal growth factor receptor. The function of this protein is unknown. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq]

Sequence: MSRPSSRAIYLHRKEYSQNLTSEPTLLQHRVEHLMTCKQGSQRVQGPEDALQKLFEMDAQGRVWSQDLILQVRDGWLQLLDIETKEELDSYRLDSIQAMNVALNTCSYNSILSITVQEPGLPGTSTLLFQCQEVGAERLKTSLQKALEEELEQSRPRLGGLQPGQDRWRGPAMERPLPMEQARYLEPGIPPEQPHQRTLEHSLPPSPRPLPRHTSAREPSAFTLPPPRRSSSPEDPERDEEVLNHVLRDIELFMGKLEKAQAKTSRKKKFGKKNKDQGGLTQAQYIDCFQKIKYSFNLLGRLATWLKETSAPELVHILFKSLNFILARCPEAGLAAQVISPLLTPKAINLLQSCLSPPESNLWMGLGPAWTTSRADWTGDEPLPYQPTFSDDWQLPEPSSQAPLGYQDPVSLRRGSHRLGSTSHFPQEKTHNHDPQPGDPNSRPSSPKPAQPALKMQVLYEFEARNPRELTVVQGEKLEVLDHSKRWWLVKNEAGRSGYIPSNILEPLQPGTPGTQGQSPSRVPMLRLSSRPEEVTDWLQAENFSTATVRTLGSLTGSQLLRIRPGELQMLCPQEAPRILSRLEAVRRMLGISP
Specifications

Specifications

AAH12926.1
Liquid
79574
EPS8L3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRPSSRAIYLHRKEYSQNLTSEPTLLQHRVEHLMTCKQGSQRVQGPEDALQKLFEMDAQGRVWSQDLILQVRDGWLQLLDIETKEELDSYRLDSIQAMNVALNTCSYNSILSITVQEPGLPGTSTLLFQCQEVGAERLKTSLQKALEEELEQSRPRLGGLQPGQDRWRGPAMERPLPMEQARYLEPGIPPEQPHQRTLEHSLPPSPRPLPRHTSAREPSAFTLPPPRRSSSPEDPERDEEVLNHVLRDIELFMGKLEKAQAKTSRKKKFGKKNKDQGGLTQAQYIDCFQKIKYSFNLLGRLATWLKETSAPELVHILFKSLNFILARCPEAGLAAQVISPLLTPKAINLLQSCLSPPESNLWMGLGPAWTTSRADWTGDEPLPYQPTFSDDWQLPEPSSQAPLGYQDPVSLRRGSHRLGSTSHFPQEKTHNHDPQPGDPNSRPSSPKPAQPALKMQVLYEFEARNPRELTVVQGEKLEVLDHSKRWWLVKNEAGRSGYIPSNILEPLQPGTPGTQGQSPSRVPMLRLSSRPEEVTDWLQAENFSTATVRTLGSLTGSQLLRIRPGELQMLCPQEAPRILSRLEAVRRMLGISP
RUO
EPS8L3
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
93.4kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPS8R3|FLJ21522|MGC16817
EPS8L3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit