Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPSTI1 Partial ORF (NP_001002264, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_001002264
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 94240
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-943-501
View Documents
Abnova™
H00094240Q01L
25 ug
Each for $769.00
Add to Cart
 
89-943-500
View Documents
Abnova™
H00094240Q01S
10 ug
Each for $499.00
Add to Cart
 
Description

Description

Sequence: MNTRNRVVNSGLGASPASRPTRDPQDPSGRQGELSPVEDQREGLEAAPKGPSRESVVHAGQRRTSAYTLIAPNINRRNEIQRIAEQELANLEKWKEQNRA
Specifications

Specifications

NP_001002264
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BRESI1/MGC29634
EPSTI1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
94240
EPSTI1 (Human) Recombinant Protein (Q01)
MNTRNRVVNSGLGASPASRPTRDPQDPSGRQGELSPVEDQREGLEAAPKGPSRESVVHAGQRRTSAYTLIAPNINRRNEIQRIAEQELANLEKWKEQNRA
RUO
EPSTI1
Wheat Germ (in vitro)
GST
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit