Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ERAF Full-length ORF (NP_057717.1, 1 a.a. - 102 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051327P01S

 View more versions of this product

Catalog No. 89-943-509


Add to cart

Description

Description

Sequence: MALLKANKDLISAGLKEFSVLLNQQVFNDPLVSEEDMVTVVEDWMNFYINYYRQQVTGEPQERDKALQELRQELNTLANPFLAKYRDFLKSHELPSHPPPSS
Specifications

Specifications

NP_057717.1
Liquid
51327
ERAF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALLKANKDLISAGLKEFSVLLNQQVFNDPLVSEEDMVTVVEDWMNFYINYYRQQVTGEPQERDKALQELRQELNTLANPFLAKYRDFLKSHELPSHPPPSS
RUO
ERAF
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AHSP|EDRF
ERAF
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit