Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ERAP2 Full-length ORF (AAH17927.1, 1 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064167P01L

 View more versions of this product

Catalog No. 89-951-056


Add to cart

Description

Description

Aminopeptidases hydrolyze N-terminal amino acids of proteins or peptide substrates. Major histocompatibility complex (MHC) class I molecules rely on aminopeptidases such as ERAP1 (MIM 606832) and LRAP to trim precursors to antigenic peptides in the endoplasmic reticulum (ER) following cleavage in the cytoplasm by tripeptidyl peptidase II (TPP2; MIM 190470) (Tanioka et al., 2003 [PubMed 12799365]).[supplied by OMIM]

Sequence: MFHSSAMVNSHRKPMFNIHRGFYCLTAILPQICICSQFSVPSSYHFTEDPGAFPVATNGERFPWQELRLPSVVIPLHYDLFVHPNLTSLDFVASEKIEVLVSNATQFIILHSKDLEITNATLQSEEDSKYMKPGKELKVLSYPAHEQIALLVPEKLTPHLKYYVAMDFQAKLGDGFEGFYKSTYRTLGGETRILAVTDFEPTQARMAFPCFDEPLFKANFSIKIRRESRHIALSNMPKVKTIELEGGLLEDHFETTVKMSTYLVAYIVCDFHSLSGFTSSGVKVSIYASPDKRNQTHYALQASLKLLDFYEKYFDIYYPLSKLGMFKFHIIVFIFAHKTCFDLFPLSLSM
Specifications

Specifications

AAH17927.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ23633|FLJ23701|FLJ23807|L-RAP|LRAP
ERAP2
Yes
wheat germ expression system
Liquid
64167
ERAP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MFHSSAMVNSHRKPMFNIHRGFYCLTAILPQICICSQFSVPSSYHFTEDPGAFPVATNGERFPWQELRLPSVVIPLHYDLFVHPNLTSLDFVASEKIEVLVSNATQFIILHSKDLEITNATLQSEEDSKYMKPGKELKVLSYPAHEQIALLVPEKLTPHLKYYVAMDFQAKLGDGFEGFYKSTYRTLGGETRILAVTDFEPTQARMAFPCFDEPLFKANFSIKIRRESRHIALSNMPKVKTIELEGGLLEDHFETTVKMSTYLVAYIVCDFHSLSGFTSSGVKVSIYASPDKRNQTHYALQASLKLLDFYEKYFDIYYPLSKLGMFKFHIIVFIFAHKTCFDLFPLSLSM
RUO
ERAP2
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit