Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ERBB2 (NP_004439.1, 676 a.a. - 1255 a.a.) Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5717

Catalog No. 89-943-522


Add to cart

Description

Description

This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 of isoform b) have been reported, with the most common allele, Ile654/Ile655, shown here. Amplification and/or overexpression of this gene has been reported in numerous cancers, including breast and ovarian tumors. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized. [provided by RefSeq]

Specifications

Specifications

NP_004439.1
Liquid
2064
ERBB2 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD340|HER-2|HER-2/neu|HER2|NEU|NGL|TKR1
ERBB2
The activity was determined by ELISA. The enzyme was incubated with biotinylated peptide in streptavidin-coated ELISA plate. Phosphorylation was detected by HRP-labeled anti-phospho antibody.
Substrate : Lyn substrate peptide. ATP: 5μM
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
67kDa
Baculovirus infected insect cell (Sf21) expression system
71% by SDS-PAGE/CBB staining.
5 ug
RUO
ERBB2
Baculovirus infected insect cell (Sf21) expression system
Insect
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit