Learn More
Abnova™ Human ERBB3 (NP_001973.2, 1 a.a - 1342 a.a) Full length Recombinant Protein with His-Flag tag at C-terminal expressed in HEK293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P6199
Catalog No. 89-943-530
Add to cart
Description
Description
This gene encodes a member of the epidermal growth factor receptor (EGFR) family of receptor tyrosine kinases. This membrane-bound protein has a neuregulin binding domain but not an active kinase domain. It therefore can bind this ligand but not convey the signal into the cell through protein phosphorylation. However, it does form heterodimers with other EGF receptor family members which do have kinase activity. Heterodimerization leads to the activation of pathways which lead to cell proliferation or differentiation. Amplification of this gene and/or overexpression of its protein have been reported in numerous cancers, including prostate, bladder, and breast tumors. Alternate transcriptional splice variants encoding different isoforms have been characterized. One isoform lacks the intermembrane region and is secreted outside the cell. This form acts to modulate the activity of the membrane-bound form. Additional splice variants have also been reported, but they have not been thoroughly characterized. [provided by RefSeq]
Specifications
Specifications
NP_001973.2
Liquid
2065
ERBB3 (Human) Recombinant Protein
Ni affinity column
SDS-PAGE (3ug/lane) stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
<1.0 EU/μg as determined by the LAL method
ERBB3
Mammalian cell (HEK293) expression system
Yes
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
72.8kDa
Mammalian cell (HEK293) expression system
>90% as determined by SEC-HPLC
25 ug
RUO
ErbB-3|HER3|LCCS2|MDA-BF-1|MGC88033|c-erbB-3|c-erbB3|erbB3-S|p180-ErbB3|p45-sErbB3|p85-sErbB3
ERBB3
Human
His-Flag