Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ERF Partial ORF (AAH22231, 181 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002077Q01S

 View more versions of this product

Catalog No. 89-943-565


Add to cart

Description

Description

Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).[supplied by OMIM]

Sequence: LGRGSVSDCSDGTSELEEPLGEDPRARPPGPPDLGAFRGPPLARLPHDPGVFRVYPRPRGGPEPLSPFPVSPLAGPGSLLPPQLSPALPMTPTHLAYTPSPTLSPMYPSG
Specifications

Specifications

AAH22231
Liquid
2077
ERF (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PE-2|PE2
ERF
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LGRGSVSDCSDGTSELEEPLGEDPRARPPGPPDLGAFRGPPLARLPHDPGVFRVYPRPRGGPEPLSPFPVSPLAGPGSLLPPQLSPALPMTPTHLAYTPSPTLSPMYPSG
RUO
ERF
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit