Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ERRFI1 Full-length ORF (NP_061821.1, 1 a.a. - 462 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054206P01S

 View more versions of this product

Catalog No. 89-943-597


Add to cart

Description

Description

ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995 [PubMed 7641805]). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000 [PubMed 10749885]; Fiorentino et al., 2000 [PubMed 11003669]).[supplied by OMIM]

Sequence: MSIAGVAAQEIRVPLKTGFLHNGRAMGNMRKTYWSSRSEFKNNFLNIDPITMAYSLNSSAQERLIPLGHASKSAPMNGHCFAENGPSQKSSLPPLLIPPSENLGPHEEDQVVCGFKKLTVNGVCASTPPLTPIKNSPSLFPCAPLCERGSRPLPPLPISEALSLDDTDCEVEFLTSSDTDFLLEDSTLSDFKYDVPGRRSFRGCGQINYAYFDTPAVSAADLSYVSDQNGGVPDPNPPPPQTHRRLRRSHSGPAGSFNKPAIRISNCCIHRASPNSDEDKPEVPPRVPIPPRPVKPDYRRWSAEVTSSTYSDEDRPPKVPPREPLSPSNSRTPSPKSLPSYLNGVMPPTQSFAPDPKYVSSKALQRQNSEGSASKVPCILPIIENGKKVSSTHYYLLPERPPYLDKYEKFFREAEETNGGAQIQPLPADCGISSATEKPDSKTKMDLGGHVKRKHLSYVVSP
Specifications

Specifications

NP_061821.1
Liquid
54206
ERRFI1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSIAGVAAQEIRVPLKTGFLHNGRAMGNMRKTYWSSRSEFKNNFLNIDPITMAYSLNSSAQERLIPLGHASKSAPMNGHCFAENGPSQKSSLPPLLIPPSENLGPHEEDQVVCGFKKLTVNGVCASTPPLTPIKNSPSLFPCAPLCERGSRPLPPLPISEALSLDDTDCEVEFLTSSDTDFLLEDSTLSDFKYDVPGRRSFRGCGQINYAYFDTPAVSAADLSYVSDQNGGVPDPNPPPPQTHRRLRRSHSGPAGSFNKPAIRISNCCIHRASPNSDEDKPEVPPRVPIPPRPVKPDYRRWSAEVTSSTYSDEDRPPKVPPREPLSPSNSRTPSPKSLPSYLNGVMPPTQSFAPDPKYVSSKALQRQNSEGSASKVPCILPIIENGKKVSSTHYYLLPERPPYLDKYEKFFREAEETNGGAQIQPLPADCGISSATEKPDSKTKMDLGGHVKRKHLSYVVSP
RUO
ERRFI1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GENE-33|MIG-6|MIG6|RALT
ERRFI1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit