Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EXOC3 Full-length ORF (NP_009208.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011336P01S

 View more versions of this product

Catalog No. 89-943-718


Add to cart

Description

Description

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. [provided by RefSeq]

Sequence: MKETDREAVATAVQRVAGMLQRPDQLDKVEQYRRREARKKASVEARLKAAIQSQLDGVRTGLSQLHNALNDVKDIQQSLADVSKDWRQSINTIESLKDVKDAVVQHSQLAAAVENLKNIFSVPEIVRETQDLIEQGALLQAHRKLMDLECSRDGLMYEQYRMDSGNTRDMTLIHGYFGSTQGLSDELAKQLWMVLQRSLVTVRRDPTLLVSVVRIIEREEKIDRRILDRKKQTGFVPPGRPKNWKEKMFTILERTVTTRIEGTQADTRESDKMWLVRHLEIIRKYVLDDLIVAKNLMVQCFPPHYEIFKNLLNMYHQALSTRMQDLASEDLEANEIVSLLTWVLNTYTSTEMMRNVELAPEVDVGTLEPLLSPHVVSELLDTYMSTLTSNIIAWLRKALETDKKDWVKETEPEADQDGYYQTTLPAIVFQMFEQNLQVAAQISEDLKTKVLVLCLQQMNSFLSRYKDEAQLYKEEHLRNRQHPHCYVQYMIAIINNCQTFKESIVSLKRKYLKNEVEEGVSPSQPSMDGILDAIAKEGCSGLLEEVFLDLEQHLNELMTKKWLLGSNAVDIICVTVEDYFNDFAKIKKPYKKRMTAEAHRRVVVEYLRAVMQKRISFRSPEERKEGAEKMVREAEQLRFLFRKLASGFGEDVDGYCDTIVAVAEVIKLTDPSLLYLEVSTLVSKYPDIRDDHIGALLAVRGDASRDMKQTIMETLEQGPAQASPSYVPLFKDIVVPSLNVAKLLK
Specifications

Specifications

NP_009208.2
Liquid
11336
EXOC3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKETDREAVATAVQRVAGMLQRPDQLDKVEQYRRREARKKASVEARLKAAIQSQLDGVRTGLSQLHNALNDVKDIQQSLADVSKDWRQSINTIESLKDVKDAVVQHSQLAAAVENLKNIFSVPEIVRETQDLIEQGALLQAHRKLMDLECSRDGLMYEQYRMDSGNTRDMTLIHGYFGSTQGLSDELAKQLWMVLQRSLVTVRRDPTLLVSVVRIIEREEKIDRRILDRKKQTGFVPPGRPKNWKEKMFTILERTVTTRIEGTQADTRESDKMWLVRHLEIIRKYVLDDLIVAKNLMVQCFPPHYEIFKNLLNMYHQALSTRMQDLASEDLEANEIVSLLTWVLNTYTSTEMMRNVELAPEVDVGTLEPLLSPHVVSELLDTYMSTLTSNIIAWLRKALETDKKDWVKETEPEADQDGYYQTTLPAIVFQMFEQNLQVAAQISEDLKTKVLVLCLQQMNSFLSRYKDEAQLYKEEHLRNRQHPHCYVQYMIAIINNCQTFKESIVSLKRKYLKNEVEEGVSPSQPSMDGILDAIAKEGCSGLLEEVFLDLEQHLNELMTKKWLLGSNAVDIICVTVEDYFNDFAKIKKPYKKRMTAEAHRRVVVEYLRAVMQKRISFRSPEERKEGAEKMVREAEQLRFLFRKLASGFGEDVDGYCDTIVAVAEVIKLTDPSLLYLEVSTLVSKYPDIRDDHIGALLAVRGDASRDMKQTIMETLEQGPAQASPSYVPLFKDIVVPSLNVAKLLK
RUO
EXOC3
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
112kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SEC6|SEC6L1|Sec6p
EXOC3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit