Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EYA4 Full-length ORF (NP_742101.2, 1 a.a. - 616 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002070P01L

 View more versions of this product

Catalog No. 89-943-779


Add to cart

Description

Description

This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal dominant hearing loss at the deafness, autosomal dominant nonsyndromic sensorineural 10 locus. Defects in this gene are also associated with dilated cardiomyopathy 1J. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MEDSQDLNEQSVKKTCTESDVSQSQNSRSMEMQDLASPHTLVGGGDTPGSSKLEKSNLSSTSVTTNGTGVSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSAHQYSPQLYPSKPYPHILSTPAAQTMSAYAGQTQYSGMQQPAVYTAYSQTGQPYSLPTYDLGVMLPAIKTESGLSQTQSPLQSGCLSYSPGFSTPQPGQTPYSYQMPGSSFAPSSTIYANNSVSNSTNFSGSQQDYPSYTAFGQNQYAQYYSASTYGAYMTSNNTADGTPSSTSTYQLQESLPGLTNQPGEFDTMQSPSTPIKDLDERTCRSSGSKSRGRGRKNNPSPPPDSDLERVFVWDLDETIIVFHSLLTGSYAQKYGKDPPMAVTLGLRMEEMIFNLADTHLFFNDLEECDQVHIDDVSSDDNGQDLSTYSFATDGFHAAASSANLCLPTGVRGGVDWMRKLAFRYRRVKELYNTYKNNVGGLLGPAKRDAWLQLRAEIEGLTDSWLTNALKSLSIISTRSNCINVLVTTTQLIPALAKVLLYSLGGAFPIENIYSATKIGKESCFERIMQRFGRKVVYVVIGDGVEEEQAAKKHNMPFWRISSHSDLLALHQALELEYL
Specifications

Specifications

NP_742101.2
Liquid
2070
EYA4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEDSQDLNEQSVKKTCTESDVSQSQNSRSMEMQDLASPHTLVGGGDTPGSSKLEKSNLSSTSVTTNGTGVSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSAHQYSPQLYPSKPYPHILSTPAAQTMSAYAGQTQYSGMQQPAVYTAYSQTGQPYSLPTYDLGVMLPAIKTESGLSQTQSPLQSGCLSYSPGFSTPQPGQTPYSYQMPGSSFAPSSTIYANNSVSNSTNFSGSQQDYPSYTAFGQNQYAQYYSASTYGAYMTSNNTADGTPSSTSTYQLQESLPGLTNQPGEFDTMQSPSTPIKDLDERTCRSSGSKSRGRGRKNNPSPPPDSDLERVFVWDLDETIIVFHSLLTGSYAQKYGKDPPMAVTLGLRMEEMIFNLADTHLFFNDLEECDQVHIDDVSSDDNGQDLSTYSFATDGFHAAASSANLCLPTGVRGGVDWMRKLAFRYRRVKELYNTYKNNVGGLLGPAKRDAWLQLRAEIEGLTDSWLTNALKSLSIISTRSNCINVLVTTTQLIPALAKVLLYSLGGAFPIENIYSATKIGKESCFERIMQRFGRKVVYVVIGDGVEEEQAAKKHNMPFWRISSHSDLLALHQALELEYL
RUO
EYA4
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
93.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CMD1J|DFNA10
EYA4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit