Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EYS Partial ORF (XP_498111, 309 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00346007Q01L

 View more versions of this product

Catalog No. 89-943-781


Add to cart

Description

Description

The product of this gene contains multiple epidermal growth factor (EGF)-like and LamG domains. The protein is expressed in the photoreceptor layer of the retina, and the gene is mutated in autosomal recessive retinitis pigmentosa. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: HVVVIQNQTLIKAYINNSLILSEDIDPHKNFVALNYDGICYLGGFEYGRKVNIVTQEIFKTNFVGKIKDVVFFQEPKNIELIKLEGYNVYDGDEQNEVT
Specifications

Specifications

XP_498111
Liquid
346007
EYS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf178|C6orf179|C6orf180|EGFL10|EGFL11|KIAA0663|RP25|SPAM|bA166P24.2|bA307F22.3|bA74E24.1|dJ1018A4.2|dJ22I17.2|dJ303F19.1
EYS
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HVVVIQNQTLIKAYINNSLILSEDIDPHKNFVALNYDGICYLGGFEYGRKVNIVTQEIFKTNFVGKIKDVVFFQEPKNIELIKLEGYNVYDGDEQNEVT
RUO
EYS
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit