Learn More
Raybiotech IncSupplier Diversity Partner Human FABP2 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHFABP21
For cell culture supernatants, plasma, and serum samples. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: In this ELISA antibody pair format, no cross-reactivity was observed with rhFABP-1, 3, 5, 6, 7, 8 and 9.
Catalog No. 50-194-9235