Learn More
Abnova™ Human FABP3 (NP_004093.1, 1 a.a. - 133 a.a.) Full length Recombinant Protein with His-Flag tag at C-terminal expressed in HEK293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P6200
Catalog No. 89-943-844
Add to cart
Description
Description
The intracellular fatty acid-binding proteins (FABPs) belongs to a multigene family. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15 kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Fatty acid-binding protein 3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is a candidate tumor suppressor gene for human breast cancer. [provided by RefSeq]
Specifications
Specifications
NP_004093.1
Liquid
2170
FABP3 (Human) Recombinant Protein
Ni affinity column
SDS-PAGE (3ug/lane) stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
<1.0 EU/μg as determined by the LAL method
FABP3
Mammalian cell (HEK293) expression system
Yes
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
18.9kDa
Mammalian cell (HEK293) expression system
>90% as determined by SEC-HPLC
25 ug
RUO
FABP11|H-FABP|MDGI|O-FABP
FABP3
Human
His-Flag