Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FAM19A4 Full-length ORF (NP_001005527.1, 1 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00151647P01S

 View more versions of this product

Catalog No. 89-943-937


Add to cart

Description

Description

This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells. Transcript variants with different 5' UTRs, but encoding the same protein, have been found for this gene. [provided by RefSeq]

Sequence: MRSPRMRVCAKSVLLSHWLFLAYVLMVCCKLMSASSQHLRGHAGHHQIKQGTCEVVAVHRCCNKNRIEERSQTVKCSCFPGQVAGTTRAQPSCVEASIVIQKWWCHMNPCLEGEDCKVLPDYSGWSCSSGNKVKTTKVTR
Specifications

Specifications

NP_001005527.1
Liquid
151647
FAM19A4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRSPRMRVCAKSVLLSHWLFLAYVLMVCCKLMSASSQHLRGHAGHHQIKQGTCEVVAVHRCCNKNRIEERSQTVKCSCFPGQVAGTTRAQPSCVEASIVIQKWWCHMNPCLEGEDCKVLPDYSGWSCSSGNKVKTTKVTR
RUO
FAM19A4
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25161|TAFA-4|TAFA4
FAM19A4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit