Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Fam40A Full-Length Orf (Bab55265.1, 1 A.A.-742 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

Manufacturer:  Abnova™ H00085369P01L

 View more versions of this product

Catalog No. 89-032-139


Add to cart

Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: MNRKCFEEDFRIHVTDKKWTELDTNQHRTHAMRLLDGLEVTAREKRLKVARAILYVAQGTFGECSSEAEVQSWMRYNIFLLLEVGTFNALVELLNMEIDNSAACSSAVRKPAISLADSTDLRVLLNIMYLIVETVHQECEGDKAEW

Specifications

Specifications

BAB55265.1
Liquid
85369
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14743|KIAA1761|MGC148091|RP4-773N10.1
FAM40A
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
FAM40A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MNRKCFEEDFRIHVTDKKWTELDTNQHRTHAMRLLDGLEVTAREKRLKVARAILYVAQGTFGECSSEAEVQSWMRYNIFLLLEVGTFNALVELLNMEIDNSAACSSAVRKPAISLADSTDLRVLLNIMYLIVETVHQECEGDKAEWRTMRQTFRAELGSPLYNNEPFAIMLFGMVTKFCSGHAPHFPMKKVLLLLWKTVLCTLGGFEELQSMKAEKRSILGLPPLPEDSIKVIRNMRAASPPASASDLIEQQQKRGRREHKALIKQDNLDAFNERDPYKADDSREEEEENDDDNSLEGETFPLERDEVMPPPLQHPQTDRLTCPKGLPWAPKVREKDIEMFLESSRSKFIGYTLGSDTNTVVGLPRPIHESIKTLKQHKYTSIAEVQAQMEEEYLRSPLSGGEEEVEQVPAETLYQGLLPSLPQYMIALLKILLAAAPTSKAKTDSINILADVLPEEMPTTVLQSMKLGVDVNRHKEVIVKAISAVLLLLLKHFKLNHVYQFEYMAQHLVFANCIPLILKFFNQNIMSYITAKNSISVLDYPHCVVHELPELTAESLEAGDSNQFCWRNLFSCINLLRILNKLTKWKHSRTMMLVVFKSAPILKRALKVKQAMMQLYVLKLLKVQTKYLGRQWRKSNMKTMSAIYQKVQHRLNDDWAYGNDLDARPWDFQAEECALRANIERFNARRYDRAHSNPDFLPVDNCLQSVLGQRVDLPEDFQMNYDLWLEREVFSKPISWEELLQ
RUO
FAM40A
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only