Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FAM44C Full-length ORF (AAH21740.1, 1 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00284257P01L

 View more versions of this product

Catalog No. 89-943-968


Add to cart

Description

Description

Sequence: MADGGGGGSGGAGPASTRASGGGGPINPASLPPGDPQLIAIIVGQLKSRGLFDSFRRDCKADVDTKPAYQNLSQKADNFVSTHLDKQEWNPPANDNQLHDGLRQSVVQSGRSEAGVDRISSQVVDPKLNHIFRPQIEQIIHEFLVAQKEAAVPALPPEPEGQDPPAPSQDTS
Specifications

Specifications

AAH21740.1
Liquid
284257
FAM44C (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADGGGGGSGGAGPASTRASGGGGPINPASLPPGDPQLIAIIVGQLKSRGLFDSFRRDCKADVDTKPAYQNLSQKADNFVSTHLDKQEWNPPANDNQLHDGLRQSVVQSGRSEAGVDRISSQVVDPKLNHIFRPQIEQIIHEFLVAQKEAAVPALPPEPEGQDPPAPSQDTS
RUO
FAM44C
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC33608
FAM44C
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit