Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human FAM49B Full-length ORF (NP_057707.3, 1 a.a. - 324 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051571P01S
Description
Sequence: MGNLLKVLTCTDLEQGPNFFLDFENAQPTESEKEIYNQVNVVLKDAEGILEDLQSYRGAGHEIREAIQHPADEKLQEKAWGAVVPLVGKLKKFYEFSQRLEAALRGLLGALTSTPYSPTQHLEREQALAKQFAEILHFTLRFDELKMTNPAIQNDFSYYRRTLSRMRINNVPAEGENEVNNELANRMSLFYAEATPMLKTLSDATTKFVSENKNLPIENTTDCLSTMASVCRVMLETPEYRSRFTNEETVSFCLRVMVGVIILYDHVHPVGAFAKTSKIDMKGCIKVLKDQPPNSVEGLLNALRYTTKHLNDETTSKQIKSMLQSpecifications
NP_057707.3 | |
Liquid | |
51571 | |
FAM49B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGNLLKVLTCTDLEQGPNFFLDFENAQPTESEKEIYNQVNVVLKDAEGILEDLQSYRGAGHEIREAIQHPADEKLQEKAWGAVVPLVGKLKKFYEFSQRLEAALRGLLGALTSTPYSPTQHLEREQALAKQFAEILHFTLRFDELKMTNPAIQNDFSYYRRTLSRMRINNVPAEGENEVNNELANRMSLFYAEATPMLKTLSDATTKFVSENKNLPIENTTDCLSTMASVCRVMLETPEYRSRFTNEETVSFCLRVMVGVIILYDHVHPVGAFAKTSKIDMKGCIKVLKDQPPNSVEGLLNALRYTTKHLNDETTSKQIKSMLQ | |
RUO | |
FAM49B | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BM-009/DKFZp686B04128/L1 | |
FAM49B | |
Yes | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title