Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FANCB Full-length ORF (NP_001018123.1, 1 a.a. - 859 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002187P01

Catalog No. 89-944-047


Add to cart

Description

Description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group B. Alternative splicing results in two transcript variants encoding the same protein. [provided by RefSeq]

Sequence: MTSKQAMSSNEQERLLCYNGEVLVFQLSKGNFADKEPTKTPILHVRRMVFDRGTKVFVQKSTGFFTIKEENSHLKIMCCNCVSDFRTGINLPYIVIEKNKKNNVFEYFLLILHSTNKFEMRLSFKLGYEMKDGLRVLNGPLILWRHVKAFFFISSQTGKVVSVSGNFSSIQWAGEIENLGMVLLGLKECCLSEEECTQEPSKSDYAIWNTKFCVYSLESQEVLSDIYIIPPAYSSVVTYVHICATEIIKNQLRISLIALTRKNQLISFQNGTPKNVCQLPFGDPCAVQLMDSGGGNLFFVVSFISNNACAVWKESFQVAAKWEKLSLVLIDDFIGSGTEQVLLLFKDSLNSDCLTSFKITDLGKINYSSEPSDCNEDDLFEDKQENRYLVVPPLETGLKVCFSSFRELRQHLLLKEKIISKSYKALINLVQGKDDNTSSAEEKECLVPLCGEEENSVHILDEKLSDNFQDSEQLVEKIWYRVIDDSLVVGVKTTSSLKLSLNDVTLSLLMDQAHDSRFRLLKCQNRVIKLSTNPFPAPYLMPCEIGLEAKRVTLTPDSKKEESFVCEHPSKKECVQIITAVTSLSPLLTFSKFCCTVLLQIMERESGNCPKDRYVVCGRVFLSLEDLSTGKYLLTFPKKKPIEHMEDLFALLAAFHKSCFQITSPGYALNSMKVWLLEHMKCEIIKEFPEVYFCERPGSFYGTLFTWKQRTPFEGILIIYSRNQTVMFQCLHNLIRILPINCFLKNLKSGSENFLIDNMAFTLEKELVTLSSLSSAIAKHESNFMQRCEVSKGKSSVVAAALSDRRENIHPYRKELQREKKKMLQTNLKVSGALYREITLKVAEVQLKSDFAAQKLSNL
Specifications

Specifications

NP_001018123.1
Liquid
2187
FANCB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSKQAMSSNEQERLLCYNGEVLVFQLSKGNFADKEPTKTPILHVRRMVFDRGTKVFVQKSTGFFTIKEENSHLKIMCCNCVSDFRTGINLPYIVIEKNKKNNVFEYFLLILHSTNKFEMRLSFKLGYEMKDGLRVLNGPLILWRHVKAFFFISSQTGKVVSVSGNFSSIQWAGEIENLGMVLLGLKECCLSEEECTQEPSKSDYAIWNTKFCVYSLESQEVLSDIYIIPPAYSSVVTYVHICATEIIKNQLRISLIALTRKNQLISFQNGTPKNVCQLPFGDPCAVQLMDSGGGNLFFVVSFISNNACAVWKESFQVAAKWEKLSLVLIDDFIGSGTEQVLLLFKDSLNSDCLTSFKITDLGKINYSSEPSDCNEDDLFEDKQENRYLVVPPLETGLKVCFSSFRELRQHLLLKEKIISKSYKALINLVQGKDDNTSSAEEKECLVPLCGEEENSVHILDEKLSDNFQDSEQLVEKIWYRVIDDSLVVGVKTTSSLKLSLNDVTLSLLMDQAHDSRFRLLKCQNRVIKLSTNPFPAPYLMPCEIGLEAKRVTLTPDSKKEESFVCEHPSKKECVQIITAVTSLSPLLTFSKFCCTVLLQIMERESGNCPKDRYVVCGRVFLSLEDLSTGKYLLTFPKKKPIEHMEDLFALLAAFHKSCFQITSPGYALNSMKVWLLEHMKCEIIKEFPEVYFCERPGSFYGTLFTWKQRTPFEGILIIYSRNQTVMFQCLHNLIRILPINCFLKNLKSGSENFLIDNMAFTLEKELVTLSSLSSAIAKHESNFMQRCEVSKGKSSVVAAALSDRRENIHPYRKELQREKKKMLQTNLKVSGALYREITLKVAEVQLKSDFAAQKLSNL
RUO
FANCB
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
124.1kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FA2|FAAP90|FAAP95|FAB|FACB
FANCB
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit