Learn More
Abnova™ Human FANCE Full-length ORF (NP_068741.1, 1 a.a. - 536 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00002178P01
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group E. [provided by RefSeq]
Sequence: MATPDAGLPGAEGVEPAPWAQLEAPARLLLQALQAGPEGARRGLGVLRALGSRGWEPFDWGRLLEALCREEPVVQGPDGRLELKPLLLRLPRICQRNLMSLLMAVRPSLPESGLLSVLQIAQQDLAPDPDAWLRALGELLRRDLGVGTSMEGASPLSERCQRQLQSLCRGLGLGGRRLKSPQAPDPEEEENRDSQQPGKRRKDSEEEAASPEGKRVPKRLRCWEEEEDHEKERPEHKSLESLADGGSASPIKDQPVMAVKTGEDGSNLDDAKGLAESLELPKAIQDQLPRLQQLLKTLEEGLEGLEDAPPVELQLLHECSPSQMDLLCAQLQLPQLSDLGLLRLCTWLLALSPDLSLSNATVLTRSLFLGRILSLTSSASRLLTTALTSFCAKYTYPVCSALLDPVLQAPGTGPAQTELLCCLVKMESLEPDAQVLMLGQILELPWKEETFLVLQSLLERQVEMTPEKFSVLMEKLCKKGLAATTSMAYAKLMLTVMTKYQANITETQRLGLAMALEPNTTFLRKSLKAALKHLGPSpecifications
NP_068741.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FACE/FAE | |
FANCE | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2178 | |
FANCE (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATPDAGLPGAEGVEPAPWAQLEAPARLLLQALQAGPEGARRGLGVLRALGSRGWEPFDWGRLLEALCREEPVVQGPDGRLELKPLLLRLPRICQRNLMSLLMAVRPSLPESGLLSVLQIAQQDLAPDPDAWLRALGELLRRDLGVGTSMEGASPLSERCQRQLQSLCRGLGLGGRRLKSPQAPDPEEEENRDSQQPGKRRKDSEEEAASPEGKRVPKRLRCWEEEEDHEKERPEHKSLESLADGGSASPIKDQPVMAVKTGEDGSNLDDAKGLAESLELPKAIQDQLPRLQQLLKTLEEGLEGLEDAPPVELQLLHECSPSQMDLLCAQLQLPQLSDLGLLRLCTWLLALSPDLSLSNATVLTRSLFLGRILSLTSSASRLLTTALTSFCAKYTYPVCSALLDPVLQAPGTGPAQTELLCCLVKMESLEPDAQVLMLGQILELPWKEETFLVLQSLLERQVEMTPEKFSVLMEKLCKKGLAATTSMAYAKLMLTVMTKYQANITETQRLGLAMALEPNTTFLRKSLKAALKHLGP | |
RUO | |
FANCE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.