Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FANCF Partial ORF (NP_073562.1, 1 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002188Q01L

 View more versions of this product

Catalog No. 89-944-056


Add to cart

Description

Description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group F. [provided by RefSeq]

Sequence: MESLLQHLDRFSELLAVSSTTYVSTWDPATVRRALQWARYLRHIHRRFGRHGPIRTALERRLHNQWRQEGGFGRGPVPGLANFQALGHCDV
Specifications

Specifications

NP_073562.1
Liquid
2188
FANCF (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FAF|MGC126856
FANCF
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MESLLQHLDRFSELLAVSSTTYVSTWDPATVRRALQWARYLRHIHRRFGRHGPIRTALERRLHNQWRQEGGFGRGPVPGLANFQALGHCDV
RUO
FANCF
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit