Learn More
Abnova™ Human FARP1 Partial ORF (NP_005757.1, 471 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010160Q01L
Description
This gene was originally isolated through subtractive hybridization due to its increased expression in differentiated chondrocytes versus dedifferentiated chondrocytes. The resulting protein contains a predicted ezrin-like domain, a Dbl homology domain, and a pleckstrin homology domain. It is believed to be a member of the band 4.1 superfamily whose members link the cytoskeleton to the cell membrane. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVSSpecifications
NP_005757.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.43kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVS | |
RUO | |
FARP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10160 | |
FARP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDEP/MGC87400/PLEKHC2 | |
FARP1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.