Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Fas Ligand/TNFSF6 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP191262
Description
The Human Fas Ligand/TNFSF6 ELISA Kit (Colorimetric) measures human Fas Ligand/TNFSF6 in cell culture supernatant, serum, plasma (edta, citrate, heparin).Specifications
0.16 - 10.0ng/mL | |
ELISA Kit | |
Human | |
356 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sFas-L, Biotin-Conjugate anti-human sFas-L monoclonal antibody, Streptavidin-HRP, Human sFas-L Standard lyophilized, 20ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Adaptive Immunity, Apoptosis, Cancer, Diabetes Research, Immunology | |
apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily |
0.07ng/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
FASLG | |
96 Tests | |
Store at 4°C. Do not freeze. | |
Fas Ligand/TNFSF6 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction