Learn More
Novus Biologicals™ Human Fas/TNFRSF6/CD95 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Supplier:  Novus Biologicals™ NBP191190
Catalog No. NBP191190
Add to Cart
Description
Description
The Human Fas/TNFRSF6/CD95 ELISA Kit (Colorimetric) measures human Fas/TNFRSF6/CD95 in cell culture supernatant, serum.
Specifications
Specifications
15.6 - 1,000pg/mL
ELISA Kit
The cross-reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human Fas/TNFRSF6/ CD95 positive sample. No cross-reactivity or interference was detected.
ELISA
FAS
96 Tests
Storage of components varies. See protocol for specific instructions.
Fas/TNFRSF6/CD95
13.2pg/mL
Cell Culture Supernatant, Serum
Human
355
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sAPO-1/Fas, Biotin-Conjugate anti-human sAPO-1/Fas monoclonal antibody, Streptavidin-HRP, Human sAPO-1/Fas Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tumor necrosis receptor superfamily member 6, TNF receptor superfamily member 6, TNFRSF6, tumor necrosis factor receptor superfamily member 6, tumor necrosis factor receptor superfamily, member 6
Product Suggestions
Product Suggestions