Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXL11 Partial ORF (NP_036440, 742 a.a. - 840 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00022992Q01S

 View more versions of this product

Catalog No. 89-944-124


Add to cart

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains at least 6 highly degenerated leucine-rich repeats. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: GAGPSDHHSASRDERFKRRQLLRLQATERTMVREKENNPSGKKELSEVEKAKIRGSYLTVTLQRPTKELHGTSIVPKLQAITASSANLRHSPRVLVQHC
Specifications

Specifications

NP_036440
Liquid
22992
FBXL11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CXXC8|DKFZp434M1735|FBL11|FBL7|FLJ00115|FLJ46431|JHDM1A|KDM2A|KIAA1004|LILINA
FBXL11
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GAGPSDHHSASRDERFKRRQLLRLQATERTMVREKENNPSGKKELSEVEKAKIRGSYLTVTLQRPTKELHGTSIVPKLQAITASSANLRHSPRVLVQHC
RUO
FBXL11
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit