Learn More
Abnova™ Human FBXL21 Partial ORF (NP_036291, 167 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026223Q01L
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains 6 tandem leucine-rich repeats. The amino acid sequence of this protein is highly similar to that of f-box and leucine-rich repeat protein 3A. Comparisons of this gene to orthologous sequences suggest that it may be a pseudogene, and may no longer express a functional protein.[provided by RefSeq]
Sequence: VSKVVLGRVGLNCPRLIELVVCANDLQPLDNELICIAEHCTNLTALGLSKCEVSCSAFIRFVRLCERRLTQLSVMEEVLIPDEDYSLDEIHTEVSKYLGRVWFPDVMPLWSpecifications
NP_036291 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VSKVVLGRVGLNCPRLIELVVCANDLQPLDNELICIAEHCTNLTALGLSKCEVSCSAFIRFVRLCERRLTQLSVMEEVLIPDEDYSLDEIHTEVSKYLGRVWFPDVMPLW | |
RUO | |
FBXL21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26223 | |
FBXL21 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBL3B/FBXL3B/FBXL3P/Fbl21/MGC120237 | |
FBXL21 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.