Learn More
Abnova™ Human FBXO15 Full-length ORF (NP_689889.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00201456P01S
Description
Members of the F-box protein family, such as FBXO15, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MPSEILLKIFSYLDAVSLLCTGCVSRRFYHLANDNFIWIGIYSTAFSPARSNWKFNSVEKIAMSMSFLSVQDKEAGYWKKEYITKQIASVKAALADILKPVNPYTGLPVKTKEALRIFGLGWAIILKEKGGKEYIMEHVDLSINDTSVTVIWYGKKWPCLASLSTLDLCGMTPVFTDWYKTPTKHRLRWHSLIAKYNLSHLTISTMIGCDRLIRIFCLHPGLLVGVWKKEEELAFVMANLHFHHLVERSTLGSATIPYELPPHSPFLDDSPEYGLHGYQLHVDLHSGGVFYLCGTFRNLFTKRGNIENGHVKLIVIHLKNNREHLPLIGKVGLSWKTDIFDGCIKSCSMMDVTLLDEHGKPFWCFSSPVCLRSPATPSDSSSFLGQTYNVDYVDAEGRVHVELVWIRETEEYLIVNLVLYLSIAKINHWFGTEYSpecifications
NP_689889.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBX15/MGC39671 | |
FBXO15 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
201456 | |
FBXO15 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPSEILLKIFSYLDAVSLLCTGCVSRRFYHLANDNFIWIGIYSTAFSPARSNWKFNSVEKIAMSMSFLSVQDKEAGYWKKEYITKQIASVKAALADILKPVNPYTGLPVKTKEALRIFGLGWAIILKEKGGKEYIMEHVDLSINDTSVTVIWYGKKWPCLASLSTLDLCGMTPVFTDWYKTPTKHRLRWHSLIAKYNLSHLTISTMIGCDRLIRIFCLHPGLLVGVWKKEEELAFVMANLHFHHLVERSTLGSATIPYELPPHSPFLDDSPEYGLHGYQLHVDLHSGGVFYLCGTFRNLFTKRGNIENGHVKLIVIHLKNNREHLPLIGKVGLSWKTDIFDGCIKSCSMMDVTLLDEHGKPFWCFSSPVCLRSPATPSDSSSFLGQTYNVDYVDAEGRVHVELVWIRETEEYLIVNLVLYLSIAKINHWFGTEY | |
RUO | |
FBXO15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.