Learn More
Abnova™ Human FBXO3 Full-length ORF (NP_036307.2, 1 a.a. - 471 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026273P02L
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants diverging at the 3' end. [provided by RefSeq]
Sequence: MAAMETETAPLTLESLPTDPLLLILSFLDYRDLINCCYVSRRLSQLSSHDPLWRRHCKKYWLISEEEKTQKNQCWKSLFIDTYSDVGRYIDHYAAIKKAWDDLKKYLEPRCPRMVLSLKEGAREEDLDAVEAQIGCKLPDDYRCSYRIHNGQKLVVPGLLGSMALSNHYRSEDLLDVDTAAGGFQQRQGLKYCLPLTFCIHTGLSQYIAVEAAEGRNKNEVFYQCPDQMARNPAAIDMFIIGATFTDWFTSYVKNVVSGGFPIIRDQIFRYVHDPECVATTGDITVSVSTSFLPELSSVHPPHYFFTYRIRIEMSKDALPEKACQLDSRYWRITNAKGDVEEVQGPGVVGEFPIISPGRVYEYTSCTTFSTTSGYMEGYYTFHFLYFKDKIFNVAIPRFHMACPTFRVSIARLEMGPDEYEEMEEEEEEEEEEDEDDDSADMDESDEDDEEERRRRVFDVPIRRRRCSRLFSpecifications
NP_036307.2 | |
Liquid | |
26273 | |
FBXO3 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAMETETAPLTLESLPTDPLLLILSFLDYRDLINCCYVSRRLSQLSSHDPLWRRHCKKYWLISEEEKTQKNQCWKSLFIDTYSDVGRYIDHYAAIKKAWDDLKKYLEPRCPRMVLSLKEGAREEDLDAVEAQIGCKLPDDYRCSYRIHNGQKLVVPGLLGSMALSNHYRSEDLLDVDTAAGGFQQRQGLKYCLPLTFCIHTGLSQYIAVEAAEGRNKNEVFYQCPDQMARNPAAIDMFIIGATFTDWFTSYVKNVVSGGFPIIRDQIFRYVHDPECVATTGDITVSVSTSFLPELSSVHPPHYFFTYRIRIEMSKDALPEKACQLDSRYWRITNAKGDVEEVQGPGVVGEFPIISPGRVYEYTSCTTFSTTSGYMEGYYTFHFLYFKDKIFNVAIPRFHMACPTFRVSIARLEMGPDEYEEMEEEEEEEEEEDEDDDSADMDESDEDDEEERRRRVFDVPIRRRRCSRLF | |
RUO | |
FBXO3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564B092/FBA/FBX3 | |
FBXO3 | |
Yes | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.