Learn More
Abnova™ Human FBXO31 Full-length ORF (AAH12748.1, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH12748.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79791 |
Molecular Weight (g/mol) | 68.1kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-944-210
|
Abnova™
H00079791P01S |
10 ug |
Each for $499.00
|
|
|||||
89-944-211
|
Abnova™
H00079791P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
Members of the F-box protein family, such as FBXO31, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MYLPPHDPHVDDPMRFKPLFRIHLMERKAATVECMYGHKGPHHGHIQIVKKDEFSTKCNQTDHHRMSGGRQEEFRTWLREEWGRTLEDIFHEHMQELILMKFIYTSQYDNCLTYRRIYLPPSRPDDLIKPGLFKGTYGSHGLEIVMLSFHGRRARGTKITGDPNIPAGQQTVEIDLRHRIQLPDLENQRNFNELSRIVLEVRERVRQEQQEGGHEAGEGRGRQGPRESQPSPAQPRAEAPSKGPDGTPGEDGGEPGDAVAAAEQPAQCGQGQPFVLPVGVSSRNEDYPRTCRMCFYGTGLIAGHGFTSPERTPGVFILFDEDRFGFVWLELKSFSLYSRVQATFRNADAPSPQAFDEMLKNIQSLTSSpecifications
AAH12748.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MYLPPHDPHVDDPMRFKPLFRIHLMERKAATVECMYGHKGPHHGHIQIVKKDEFSTKCNQTDHHRMSGGRQEEFRTWLREEWGRTLEDIFHEHMQELILMKFIYTSQYDNCLTYRRIYLPPSRPDDLIKPGLFKGTYGSHGLEIVMLSFHGRRARGTKITGDPNIPAGQQTVEIDLRHRIQLPDLENQRNFNELSRIVLEVRERVRQEQQEGGHEAGEGRGRQGPRESQPSPAQPRAEAPSKGPDGTPGEDGGEPGDAVAAAEQPAQCGQGQPFVLPVGVSSRNEDYPRTCRMCFYGTGLIAGHGFTSPERTPGVFILFDEDRFGFVWLELKSFSLYSRVQATFRNADAPSPQAFDEMLKNIQSLTS | |
RUO | |
FBXO31 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79791 | |
FBXO31 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434B027/DKFZp434J1815/FBX14/FBXO14/FLJ22477/Fbx31/MGC15419/MGC9527/pp2386 | |
FBXO31 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.