Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXO36 Full-length ORF (AAH33935.1, 1 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00130888P01S

 View more versions of this product

Catalog No. 89-944-225


Add to cart

Description

Description

Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MASWLPETLFETVGQGPPPSKDYYQLLVTRSQVIFRWWKISLRSEYRSTKPGEAKETHEDFLENSHLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTNKLQLQRQLRKRKQKYGNLREKQP
Specifications

Specifications

AAH33935.1
Liquid
130888
FBXO36 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASWLPETLFETVGQGPPPSKDYYQLLVTRSQVIFRWWKISLRSEYRSTKPGEAKETHEDFLENSHLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTNKLQLQRQLRKRKQKYGNLREKQP
RUO
FBXO36
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37592|FLJ41090|Fbx36
FBXO36
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit