Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXO46 Partial ORF (XP_371179, 176 a.a. - 275 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023403Q01L

 View more versions of this product

Catalog No. 89-944-252


Add to cart

Description

Description

Members of the F-box protein family, such as FBXO46, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: VALVEQRAALALQSYPRPTTPAPVVFVSAEQGGPAKGVGSERRSGGGDCSRVAEAVAHFEAQRDSPPTKGLRKEERPGPGPGEVRIAFRISNGREPRAPD
Specifications

Specifications

XP_371179
Liquid
23403
FBXO46 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
20D7-FC4|FBXO34L|Fbx46
FBXO46
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VALVEQRAALALQSYPRPTTPAPVVFVSAEQGGPAKGVGSERRSGGGDCSRVAEAVAHFEAQRDSPPTKGLRKEERPGPGPGEVRIAFRISNGREPRAPD
RUO
FBXO46
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit