Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXO6 Full-length ORF (AAH20880.1, 1 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026270P01S

 View more versions of this product

Catalog No. 89-944-257


Add to cart

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle. [provided by RefSeq]

Sequence: MDAPHSKAALDSINELPENILLELFTHVPARQLLLNCRLVCSLWRDLIDLMTLWKRKCLREGFITKDWDQPVADWKIFYFLRSLHRNLLRNPCAEEDMFAWQIDFNGGDRWKVESLPGAHGTDFPDPKVKKYFVTSYEMCLKSQLVDLVAEGYWEELLDTFRPDIVVKDWFAARADCGCTYQLKVQLASADYFVLASFEPPPVTIQQWNNATWTEVSYTFSDYPRGVRYILFQHGGRDTQYWAGWYGPRVTNSSIVVSPKMTRNQASSEAQPGQKHGQEEAAQSPYRAVVQIF
Specifications

Specifications

AAH20880.1
Liquid
26270
FBXO6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDAPHSKAALDSINELPENILLELFTHVPARQLLLNCRLVCSLWRDLIDLMTLWKRKCLREGFITKDWDQPVADWKIFYFLRSLHRNLLRNPCAEEDMFAWQIDFNGGDRWKVESLPGAHGTDFPDPKVKKYFVTSYEMCLKSQLVDLVAEGYWEELLDTFRPDIVVKDWFAARADCGCTYQLKVQLASADYFVLASFEPPPVTIQQWNNATWTEVSYTFSDYPRGVRYILFQHGGRDTQYWAGWYGPRVTNSSIVVSPKMTRNQASSEAQPGQKHGQEEAAQSPYRAVVQIF
RUO
FBXO6
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.97kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBG2|FBS2|FBX6|Fbx6b
FBXO6
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit